Learn More
New England Biolabs, Inc. NEBNext Ultra™ II RNA First Strand Synthesis Module – 96 reactions
SDP
Catalog No. 501526377
Encompass
This module is part of the Ultra II Directional RNA and Ultra II RNA workflows, for Illumina- compatible RNA library construction. These workflows are compatible with poly(A) mRNA isolation or ribosomal RNA depletion, and enable high yield preparation of high quality libraries from 5 ng - 1 g total RNA. The NEBNext Ultra II RNA First Strand Synthesis Module has been optimized to convert a broad range of input amounts of RNA into cDNA using random priming. The module is optimized for use with the NEBNext Ultra II Directional RNA Second Strand Synthesis Module or the NEBNext Ultra II Non-Directional RNA Second Strand Synthesis Module, and is part of the Ultra II RNA directional or non-directional workflows. These workflows are compatible with poly(A) mRNA isolation or ribosomal RNA depletion, and enable high yield preparation of high quality libraries from 5 ng - 1µg total RNA.
Catalog No. 50-152-6377 Supplier New England Biolabs, Inc. Supplier No. E7771L
This module is part of the Ultra II Directional RNA and Ultra II RNA workflows, for Illumina- compatible RNA library construction. These workflows are compatible with poly(A) mRNA isolation or ribosomal RNA depletion, and enable high yield preparation of high quality libraries from 5 ng - 1 g total RNA. The NEBNext Ultra II RNA First Strand Synthesis Module has been optimized to convert a broad range of input amounts of RNA into cDNA using random priming. The module is optimized for use with the NEBNext Ultra II Directional RNA Second Strand Synthesis Module or the NEBNext Ultra II Non-Directional RNA Second Strand Synthesis Module, and is part of the Ultra II RNA directional or non-directional workflows. These workflows are compatible with poly(A) mRNA isolation or ribosomal RNA depletion, and enable high yield preparation of high quality libraries from 5 ng - 1µg total RNA.