Learn More
New England Biolabs, Inc. NEBNext Ultra™ II Directional RNA Second Strand Synthesis Module – 96 reactions
SDP
Catalog No. 501526367
Encompass
The NEBNext Ultra II Directional RNA Second Strand Synthesis Module has been optimized to generate double-stranded cDNA from first-strand cDNA, as part of the NEBNext Directional RNA library preparation workflow. This workflow uses the dUTP method for strand-specific library construction, which depends on incorporation of uracil into the second strand cDNA, enabled by the NEBNext Second Strand Synthesis Reaction Buffer with dUTP Mix, included in this module.
Catalog No. 50-152-6367 Supplier New England Biolabs, Inc. Supplier No. E7550L
The NEBNext Ultra II Directional RNA Second Strand Synthesis Module has been optimized to generate double-stranded cDNA from first-strand cDNA, as part of the NEBNext Directional RNA library preparation workflow. This workflow uses the dUTP method for strand-specific library construction, which depends on incorporation of uracil into the second strand cDNA, enabled by the NEBNext Second Strand Synthesis Reaction Buffer with dUTP Mix, included in this module.