Learn More
Charles River Laboratories Endosafe nexgen-MCS™ instrument
Catalog No. NC1411380
Encompass
The Endosafe nexgen-MCS endotoxin testing benchtop system is comprised of five individual spectrophotometers built into a unit with a single Ethernet connector that links to your computer running EndoScan-V™ version 5.5.5 or higher, the 21 CFR Part 11 compliant-ready endotoxin measuring software.
Catalog No. NC1411380 Supplier Charles River Laboratories Supplier No. MCS150K
Add to Cart
Add to Cart
Request a Quote
The Endosafe nexgen-MCS endotoxin testing benchtop system is comprised of five individual spectrophotometers built into a unit with a single Ethernet connector that links to your computer running EndoScan-V™ version 5.5.5 or higher, the 21 CFR Part 11 compliant-ready endotoxin measuring software.