Learn More
Perkin Elmer US LLC NexION 1000/2000 Cell Stability Solution Without Se, 1% HNO3, 500 mL
Catalog No. 502113692
Encompass_Preferred
Concentration of Element includes: Matrix Concentration Volume 1 % HNO3 1 µg/L: Cd, Cr, Fe, Mg, Pb 500 mL 10 µg/L: Co, Cu
Catalog No. 50-211-3692 Supplier Perkin Elmer US LLC Supplier No. N8151033
Concentration of Element includes: Matrix Concentration Volume 1 % HNO3 1 µg/L: Cd, Cr, Fe, Mg, Pb 500 mL 10 µg/L: Co, Cu