Learn More
Perkin Elmer US LLC NexION 5000 Stability Solution 1% HNO3, 500 mL
Catalog No. 502117651
Encompass_Preferred
Concentration of Element includes: Element Concentration Cadmium 200 µg/mL Chromium 200 µg/mL Iron 200 µg/mL Lithium 200 µg/mL Magnesium 200 µg/mL Lead 200 µg/mL Copper 1 µg/mL Nitric Acid 1%
Catalog No. 50-211-7651 Supplier Perkin Elmer US LLC Supplier No. N8162302
Concentration of Element includes: Element Concentration Cadmium 200 µg/mL Chromium 200 µg/mL Iron 200 µg/mL Lithium 200 µg/mL Magnesium 200 µg/mL Lead 200 µg/mL Copper 1 µg/mL Nitric Acid 1%