Learn More
Perkin Elmer US LLC NexION AFT Multi-Element Solution (H/He), 2% HNO3, 125 mL
Catalog No. 502104396
Encompass_Preferred
Concentration of Element includes: Matrix Concentration Volume 2 % HNO3 10 µg/L: Ag, Al, As, Ba, Be, Bi, Ca, Cd, Co, Cr, Cs, Cu, Fe, Ga, ln, K, Li, Mg, Mn, Na, Ni, Pb, Rb, Se, Sr, Tl, U, V, Zn 125 mL
Catalog No. 50-210-4396 Supplier Perkin Elmer US LLC Supplier No. N8151038
Concentration of Element includes: Matrix Concentration Volume 2 % HNO3 10 µg/L: Ag, Al, As, Ba, Be, Bi, Ca, Cd, Co, Cr, Cs, Cu, Fe, Ga, ln, K, Li, Mg, Mn, Na, Ni, Pb, Rb, Se, Sr, Tl, U, V, Zn 125 mL