Learn More
Perkin Elmer US LLC NexION 350X/D/S/1000/2000 Cell Stability Solution, 1% HNO3, 500 mL
Catalog No. NC0879892
Encompass_Preferred
Concentration of Element includes: Matrix Concentration Volume 1 % HNO3 1 µg/L: Cd, Cr, Fe, Mg, Pb 500 mL 10 µg/L: Co, Cu, ln, Se
Catalog No. NC0879892 Supplier Perkin Elmer US LLC Supplier No. N8145054
Concentration of Element includes: Matrix Concentration Volume 1 % HNO3 1 µg/L: Cd, Cr, Fe, Mg, Pb 500 mL 10 µg/L: Co, Cu, ln, Se