Learn More
Perkin Elmer US LLC NexION DRC Mode Detection Limit Blank Solution, 0.5 % HNO3, 100 mL
Catalog No. 502104847
Encompass_Preferred
Concentration of Element includes: Matrix Concentration 0.5 % HNO3 <1 µg/L: Si, P < 1 µg/L: B, Na, K < 0.2 µg/L: Zn, Al < 0.1 µg/L: All other metals and non-metals
Catalog No. 50-210-4847 Supplier Perkin Elmer US LLC Supplier No. N8145055
Add to Cart
Add to Cart
Concentration of Element includes: Matrix Concentration 0.5 % HNO3 <1 µg/L: Si, P < 1 µg/L: B, Na, K < 0.2 µg/L: Zn, Al < 0.1 µg/L: All other metals and non-metals