Learn More
Perkin Elmer US LLC NexION DRC Mode Detection Limit Solution, 0.5 % HNO3, 100 mL
Catalog No. 502104846
Encompass_Preferred
Concentration of Element includes: Matrix Concentration Volume 0.5 % HNO3 1 µg/L: Be, Ca, Co, Fe, ln, U 100 mL
Catalog No. 50-210-4846 Supplier Perkin Elmer US LLC Supplier No. N8145056
Concentration of Element includes: Matrix Concentration Volume 0.5 % HNO3 1 µg/L: Be, Ca, Co, Fe, ln, U 100 mL