Learn More
Perkin Elmer US LLC NexION Dual Detector Solution, 2% HNO3, 100 mL
Catalog No. NC0543461
Encompass_Preferred
Concentration of Element includes: Matrix Concentration Volume 2% HNO3 200 µg/L: Al, Ba, Ce, Co, Cu, ln, Li, Mg, Mn, Ni, Pb, Tb, U, Zn 100 mL
Catalog No. NC0543461 Supplier Perkin Elmer US LLC Supplier No. N8145059
Concentration of Element includes: Matrix Concentration Volume 2% HNO3 200 µg/L: Al, Ba, Ce, Co, Cu, ln, Li, Mg, Mn, Ni, Pb, Tb, U, Zn 100 mL