Learn More
Perkin Elmer US LLC NexION KED Mode Detection Limit Solution, 1 % HCl, 100 mL
Catalog No. 502104839
Encompass_Preferred
Concentration of Element includes: Matrix Concentration Volume 1 % HCl 10 µg/L: V, As, Se 100 mL
Catalog No. 50-210-4839 Supplier Perkin Elmer US LLC Supplier No. N8145058
Concentration of Element includes: Matrix Concentration Volume 1 % HCl 10 µg/L: V, As, Se 100 mL