Learn More
Perkin Elmer US LLC NexION KED Mode Detection Limit Blank Solution, 1 % HCl, 100 mL
Catalog No. 502104840
Encompass_Preferred
Concentration of Element includes: Matrix Concentration 1 % HNO3 <1 µg/L: Si, P < 1 µg/L: B, Na, K < 0.2 µg/L: Zn, Al, Ca, Fe < 0.1 µg/L: All other metals and non-metals
Catalog No. 50-210-4840 Supplier Perkin Elmer US LLC Supplier No. N8145057
Concentration of Element includes: Matrix Concentration 1 % HNO3 <1 µg/L: Si, P < 1 µg/L: B, Na, K < 0.2 µg/L: Zn, Al, Ca, Fe < 0.1 µg/L: All other metals and non-metals