Learn More
Perkin Elmer US LLC NexION KED Setup Solution, 1 %, HNO3, 500 mL
Catalog No. 502117278
Encompass_Preferred
Concentration of Element includes: Matrix Concentration Volume 1% HNO3 10 µg/mL: Be, Ce, Fe, ln, Li, Mg, Pb, U 500 mL
Catalog No. 50-211-7278 Supplier Perkin Elmer US LLC Supplier No. N8145289
Concentration of Element includes: Matrix Concentration Volume 1% HNO3 10 µg/mL: Be, Ce, Fe, ln, Li, Mg, Pb, U 500 mL