Learn More
Perkin Elmer US LLC Demountable Sapphire Injector, 1.8 mm I.D.
Catalog No. NC1984925
Encompass_Preferred
This Sapphire Injector Cassette Mount is designed for use with demountable torches in NexION 300/350 ICP-MS series instruments. The demountable design allows the injector to be removed and replaced efficiently without dismounting the torch. Standard Injectors have an O-ring free direct connection to Cyclonic and PFA spray chambers while O-ring free Injectors have an O-ring free seal that reduces contamination, makes it easy to clean and easy to insert.
Catalog No. NC1984925 Supplier Perkin Elmer US LLC Supplier No. N8122411
This Sapphire Injector Cassette Mount is designed for use with demountable torches in NexION 300/350 ICP-MS series instruments. The demountable design allows the injector to be removed and replaced efficiently without dismounting the torch. Standard Injectors have an O-ring free direct connection to Cyclonic and PFA spray chambers while O-ring free Injectors have an O-ring free seal that reduces contamination, makes it easy to clean and easy to insert.