Learn More
PREMIER BIOTECH Nexscreen 10-Panel Cup 25/Cs
Catalog No. NC2002054
Encompass_Preferred
AMP500/BAR300/BZO300/COC150/MDMA500/MET500/MTD300/OPI2000/OXY100/THC50.  NexScreen Cup, integrated rapid drug test cup, set the standard for advanced rapid drug testing. It is accurate, easy, rugged, and flexible. Accurate: The NexScreen Cup (FDA-cleared, CLIA-waived, CE marked) provides the same accuracy as a laboratory. In an independent study, the NexScreen Cup provided 100% correlation at 25% and 50% above and below cutoff. NexScreen Cup¿s testing accuracy has been published in the Journal of Analytical Toxicology and endorsed by the Society of Forensic Toxicologists. Simplicity: Just collect the specimen, peel away the label, and read the color-coded results. There¿s only 1 drug test on each strip making it easy to read.
This item is not returnable. View return policy
Catalog No. NC2002054 Supplier PREMIER BIOTECH Supplier No. ND61012N
AMP500/BAR300/BZO300/COC150/MDMA500/MET500/MTD300/OPI2000/OXY100/THC50.  NexScreen Cup, integrated rapid drug test cup, set the standard for advanced rapid drug testing. It is accurate, easy, rugged, and flexible. Accurate: The NexScreen Cup (FDA-cleared, CLIA-waived, CE marked) provides the same accuracy as a laboratory. In an independent study, the NexScreen Cup provided 100% correlation at 25% and 50% above and below cutoff. NexScreen Cup¿s testing accuracy has been published in the Journal of Analytical Toxicology and endorsed by the Society of Forensic Toxicologists. Simplicity: Just collect the specimen, peel away the label, and read the color-coded results. There¿s only 1 drug test on each strip making it easy to read.