Learn More
New England Biolabs, Inc. NEBNext dsDNA Fragmentase – 250 reactions
SDP
Catalog No. 50591108
Encompass
NEBNext dsDNA Fragmentase is an enzyme-based reagent that shears DNA to produce fragments of the desired sizes in a time-dependent manner, for next generation sequencing library preparation protocols Note: NEBNext dsDNA Fragmentase is not part of the NEBNext Ultra II FS (Fragmentation System) reagents.
Catalog No. 50-591-108 Supplier New England Biolabs, Inc. Supplier No. M0348L
NEBNext dsDNA Fragmentase is an enzyme-based reagent that shears DNA to produce fragments of the desired sizes in a time-dependent manner, for next generation sequencing library preparation protocols Note: NEBNext dsDNA Fragmentase is not part of the NEBNext Ultra II FS (Fragmentation System) reagents.