Learn More
Bioworld Next Gel Running Buffer (20X), 500 mL
Catalog No. 50198640
Encompass_Preferred
Product DescriptionNext Gelrunning buffer has a concentration of 20X and a pH level at 25°C between 7.4 – 8.0. Proteomics grade.
Catalog No. 50-198-640 Supplier Bioworld Supplier No. 214200172
Product DescriptionNext Gelrunning buffer has a concentration of 20X and a pH level at 25°C between 7.4 – 8.0. Proteomics grade.