Learn More
SCHOTT NORTH AMERICA INC Nexterion Slide A+ – Aminosilane Coated, 25/pack
SDP
Catalog No. NC2165913
Encompass
This item has a minimum order quantity of 2 packs per supplier requirementsCleanroom cleaned and packaged. Slide dimensions:75.6 x 25.0 x 1.0 mm
This item is not returnable. View return policy
Catalog No. NC2165913 Supplier SCHOTT NORTH AMERICA INC Supplier No. 1064875
This item has a minimum order quantity of 2 packs per supplier requirementsCleanroom cleaned and packaged. Slide dimensions:75.6 x 25.0 x 1.0 mm