Learn More
SCHOTT NORTH AMERICA INC Nexterion Slide AL- Aldehydesilane Coated 25/pack
SDP
Catalog No. NC2145598
Encompass
This item has a minimum order quantity of 2 packs per supplier requirementsCleanroom cleaned and packaged. Slide dimensions:75.6 x 25.0 x 1.0 mm
Catalog No. NC2145598 Supplier SCHOTT NORTH AMERICA INC Supplier No. 1064874
This item has a minimum order quantity of 2 packs per supplier requirementsCleanroom cleaned and packaged. Slide dimensions:75.6 x 25.0 x 1.0 mm