Learn More
Alkali Scientific NextPette™ Pipettor, 20 to 200ul
SDP
Catalog No. 5000663085
Encompass_Preferred
NextPette's materials and components were selected to minimize while ensuring durability and long service life. The pipettors are designed with service in mind, allowing easy disassembly and simple seal replacement. Silicone lubricant is included with each pipettor. Recalibration can be performed in the lab without tools, and the instruction manual explains how to check pipetting accuracy and recalibrate when required. NextPette™ variable volume pipette 0.1 to 1.0µL features an ergonomic design ideal for high volume laboratories. Adjust volume easily with one hand while viewing the large 4 digit display. Comfortable stroke distance suits all hand s. Individually tested for accuracy according to EN ISO 8655 and sold individually. Bulk Bag 250/pack for P9700 T5M and 100/pack for P9700 T10M. Macro Set P9700 MARCO includes P9700 20 (20–200ul), P9700 1000 (100–1000ul), and P9700 5M (1–5ml).
Catalog No. 50-006-63085 Supplier Alkali Scientific Supplier No. P9700200
NextPette's materials and components were selected to minimize while ensuring durability and long service life. The pipettors are designed with service in mind, allowing easy disassembly and simple seal replacement. Silicone lubricant is included with each pipettor. Recalibration can be performed in the lab without tools, and the instruction manual explains how to check pipetting accuracy and recalibrate when required. NextPette™ variable volume pipette 0.1 to 1.0µL features an ergonomic design ideal for high volume laboratories. Adjust volume easily with one hand while viewing the large 4 digit display. Comfortable stroke distance suits all hand s. Individually tested for accuracy according to EN ISO 8655 and sold individually. Bulk Bag 250/pack for P9700 T5M and 100/pack for P9700 T10M. Macro Set P9700 MARCO includes P9700 20 (20–200ul), P9700 1000 (100–1000ul), and P9700 5M (1–5ml).