Learn More
Alkali Scientific NextPette™ Pipettor, Macro Set includes: P9700 20 (20 to 200ul), P9700 1000 (100 to 1000ul), P9700 5M (1 to 5ml), P9700 10M (1 to 10ml), P9700 CAR (Carousel).
SDP
Catalog No. 5000662123
Encompass_Preferred
NextPette's materials and components were selected to minimize while ensuring durability and long service life. The pipettors are designed for easy servicing and are simple to disassemble, with seals that are easy to replace. Silicone lubricant is included with each pipettor. Recalibration can be performed in the lab without tools, and the manual provides instructions for checking pipetting accuracy and recalibrating when needed. NextPette™ variable volume pipette 0.1 to 1.0µL features an ergonomic design for high volume laboratories. Adjust volume easily with one hand while viewing the large 4 digit display. Comfortable stroke distance suits all hand s. Individually tested for accuracy according to EN ISO 8655 and sold individually. Bulk Bag 250/pack for P9700 T5M and 100/pack for P9700 T10M. Macro Set P9700 MARCO includes P9700 20 (20–200ul), P9700 1000 (100–1000ul), and P9700 5M (1–5ml).
Catalog No. 50-006-62123 Supplier Alkali Scientific Supplier No. P9700MARCO
NextPette's materials and components were selected to minimize while ensuring durability and long service life. The pipettors are designed for easy servicing and are simple to disassemble, with seals that are easy to replace. Silicone lubricant is included with each pipettor. Recalibration can be performed in the lab without tools, and the manual provides instructions for checking pipetting accuracy and recalibrating when needed. NextPette™ variable volume pipette 0.1 to 1.0µL features an ergonomic design for high volume laboratories. Adjust volume easily with one hand while viewing the large 4 digit display. Comfortable stroke distance suits all hand s. Individually tested for accuracy according to EN ISO 8655 and sold individually. Bulk Bag 250/pack for P9700 T5M and 100/pack for P9700 T10M. Macro Set P9700 MARCO includes P9700 20 (20–200ul), P9700 1000 (100–1000ul), and P9700 5M (1–5ml).