Learn More
Alkali Scientific NextPette™ Pipettor, MicroSet includes: P9700 10 (0.5 to 10ul), P9700 20 (2 to 20ul), P9700 200 (20 to 200ul), P9700 1000 (100 to 1000ul), P9700 CAR (Carousel).
SDP
Catalog No. 5000662115
Encompass_Preferred
NextPette's materials and components were carefully selected to minimize and ensure durability and a long service life. The pipettors have also been designed with service in mind. They are easy to disassemble, and seals are easy to replace. Silicone lubricant is included with each pipettor. Recalibration can be done in the lab no tools are required and the instruction manual includes full details on how to properly check pipetting accuracy and how to recalibrate if necessary. NextPette™ variable volume pipette 0.1 to 1.0µL Ergonomic design is ideal for use in high volume laboratories Adjust volume easily with just one hand, while still being able to see the large 4 digit display Comfortable stroke distance for all hand s Individually tested for accuracy according to EN ISO 8655 Sold individually Bulk Bag 250/pack for P9700 T5M, and 100/pack for P9700 T10M Macro Set P9700 MARCO Includes: P9700 20 (20 to 200ul), P9700 1000 (100 to 1000ul), P9700 5M (1 to 5ml), P9700 10M (1
Catalog No. 50-006-62115 Supplier Alkali Scientific Supplier No. P9700MICRO
NextPette's materials and components were carefully selected to minimize and ensure durability and a long service life. The pipettors have also been designed with service in mind. They are easy to disassemble, and seals are easy to replace. Silicone lubricant is included with each pipettor. Recalibration can be done in the lab no tools are required and the instruction manual includes full details on how to properly check pipetting accuracy and how to recalibrate if necessary. NextPette™ variable volume pipette 0.1 to 1.0µL Ergonomic design is ideal for use in high volume laboratories Adjust volume easily with just one hand, while still being able to see the large 4 digit display Comfortable stroke distance for all hand s Individually tested for accuracy according to EN ISO 8655 Sold individually Bulk Bag 250/pack for P9700 T5M, and 100/pack for P9700 T10M Macro Set P9700 MARCO Includes: P9700 20 (20 to 200ul), P9700 1000 (100 to 1000ul), P9700 5M (1 to 5ml), P9700 10M (1