Learn More
Selleck Chemical LLC Nexturastat A
SDP
Catalog No. 5000003678
Encompass_Preferred
Nexturastat A is a potent and selective HDAC6 inhibitor with IC50 of 5 nM 190-fold selectivity over other HDACs
Catalog No. 50-000-03678 Supplier Selleck Chemical LLC Supplier No. S74735MG
Nexturastat A is a potent and selective HDAC6 inhibitor with IC50 of 5 nM 190-fold selectivity over other HDACs