Learn More
Apexbio Technology LLC Nexturastat A 1403783-31-2 10mM (in 1mL DMSO)
SDP
Catalog No. 5000063084
Encompass_Preferred
Nexturastat A (CAS 1403783-31-2) is a selective small-molecule inhibitor of histone deacetylase 6 (HDAC6) It displays potent inhibitory activity against HDAC6 with an IC50 of 5 2 nM While predominantly targeting HDAC6 Nexturastat A also exhibits moderate inhibitory effects on other HDAC isoforms (HDAC1-5 and 7-11) at low micromolar concentrations In cell-based studies including mouse B16 melanoma cell lines Nexturastat A has shown antiproliferative properties and induction of apoptosis Due to its selective HDAC6 inhibition Nexturastat A is utilized in cancer research focusing on epigenetic regulation and therapeutic development
Catalog No. 50-000-63084 Supplier Apexbio Technology LLC Supplier No. B37045.1
Nexturastat A (CAS 1403783-31-2) is a selective small-molecule inhibitor of histone deacetylase 6 (HDAC6) It displays potent inhibitory activity against HDAC6 with an IC50 of 5 2 nM While predominantly targeting HDAC6 Nexturastat A also exhibits moderate inhibitory effects on other HDAC isoforms (HDAC1-5 and 7-11) at low micromolar concentrations In cell-based studies including mouse B16 melanoma cell lines Nexturastat A has shown antiproliferative properties and induction of apoptosis Due to its selective HDAC6 inhibition Nexturastat A is utilized in cancer research focusing on epigenetic regulation and therapeutic development