Learn More
Medchemexpress LLC HY-103708 10mg Medchemexpress, Nezukone CAS:13656-81-0 Purity:>98%
SDP
Catalog No. 501968169
Encompass_Preferred
Medchemexpress, HY-103708 10mg Nezukone CAS:13656-81-0 Nezukone is a derivative of hinokitiol. Hinokitiol can restore iron transports, however, Nezukone cannot bind or transport iron. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-8169 Supplier Medchemexpress LLC Supplier No. HY10370810MG
Medchemexpress, HY-103708 10mg Nezukone CAS:13656-81-0 Nezukone is a derivative of hinokitiol. Hinokitiol can restore iron transports, however, Nezukone cannot bind or transport iron. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.