Learn More
TARGETMOL CHEMICALS INC NF-56-EJ40 5MG
SDP
Catalog No. 502581477
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. NF-56-EJ40 is an effective and highly selective antagonist of human SUCNR1 with an IC50 of 25 nM and a Ki of 33 nM. purity: 98%
Catalog No. 50-258-1477 Supplier TARGETMOL CHEMICALS INC Supplier No. T122165MG
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. NF-56-EJ40 is an effective and highly selective antagonist of human SUCNR1 with an IC50 of 25 nM and a Ki of 33 nM. purity: 98%