Learn More
Medchemexpress LLC HY-134476 5mg Medchemexpress, NF-κΒ activator 1 CAS:2387524-59-4 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1344765MG
Encompass_Preferred
Medchemexpress, HY-134476 5mg NF-κΒ activator 1 CAS:2387524-59-4 NF-κΒ activator 1 is a potent NF-κΒ activator with an EC50 of 0.9 μM. NF-κΒ activator 1 induces superoxide dismutase (SOD)2 mRNA expression. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-1367