Learn More
Enzo Life Sciences NF-ATc1 monoclonal antibody (7A6) (100µl)
SDP
Supplier:  Enzo Life Sciences ALX804022R100
Encompass_Preferred
Clone: 7A6. Isotype: IgG1. Immunogen: Recombinant human NF-ATc1 (nuclear factor of activated T cells, cytoplasmic 1) (aa 1-654). Species reactivity: Human, Mouse, Rat, Monkey. Applications: ICC, IF, IHC, IP, WB Gel Shift, ChIP. Formulation: Liquid. Ascites diluted in PBS containing 0.05% sodium azide.
Catalog No. 50-200-9751