Learn More
Enzo Life Sciences NF-ATc2 monoclonal antibody (25A10.D6.D2) (100µg)
SDP
Supplier:  Enzo Life Sciences ALX804215C100
Encompass_Preferred
Clone: 25A10.D6.D2. Isotype: IgG1. Immunogen: Synthetic peptide corresponding to aa 51-69 (A51ISSPSGLAYPDDVLDYGL69) of mouse nuclear factor of activated T cells, cytoplasmic 2 (NF-ATc2) conserved in all three isoforms A, B and C. Species reactivity: Human, Mouse, Rat. Applications: ICC, IF, IHC (FS), IHC (PS), IP, WB Gel Supershift. Formulation: Liquid. Purified antibody in PBS containing 0.05% sodium azide.
Catalog No. 50-200-9832