Learn More
BIOHIPPO NF B-RFP-Luc-BSD LV
SDP
Catalog No. 5000330729
Encompass
NF B-RFP-Luc-BSD is a transcription factor reporter lentiviral vector designed to monitor NF B signaling pathway in living cells The construct carries a RFP and luciferase dual reporter under the control of pathway responsive elements and includes a blasticidin selection marker to generate stable cell pools The lentivirus is provided as a ready to use VSV G pseudotyped preparation for efficient gene delivery and stable genomic integration Each vial provides a nominal titer of 5x10E6 transducing units TU Typical applications include pathway activation studies assay development compound screening and mechanism of action research The system is suitable for human and mouse cells and can be read using standard fluorescence microscopy flow cytometry or luciferase assays depending on the reporter gene This product is intended for research use only and is not for diagnostic or therapeutic procedures
Catalog No. 50-003-30729 Supplier BIOHIPPO Supplier No. BHV1940000212
NF B-RFP-Luc-BSD is a transcription factor reporter lentiviral vector designed to monitor NF B signaling pathway in living cells The construct carries a RFP and luciferase dual reporter under the control of pathway responsive elements and includes a blasticidin selection marker to generate stable cell pools The lentivirus is provided as a ready to use VSV G pseudotyped preparation for efficient gene delivery and stable genomic integration Each vial provides a nominal titer of 5x10E6 transducing units TU Typical applications include pathway activation studies assay development compound screening and mechanism of action research The system is suitable for human and mouse cells and can be read using standard fluorescence microscopy flow cytometry or luciferase assays depending on the reporter gene This product is intended for research use only and is not for diagnostic or therapeutic procedures