Learn More
Enzo Life Sciences Neurofilaments (phosphorylated) monoclonal antibody (SMI31) (100µl)
SDP
Supplier:  Enzo Life Sciences ENZABS2150100
Encompass_Preferred
Clone: SMI-31. Isotype: IgG1. Species reactivity: Human Xenopus, Mammalian, Chicken. Specificity: Recognizes a phosphorylated epitope in extensively phosphorylated neurofilament H and, to a lesser extent, in neurofilament M. Applications: ELISA, ICC, IHC, WB. Purity: Purified. Formulation: Liquid. In PBS containing thimerosal.
Catalog No. 50-201-2764