Learn More
Cell Signaling Technology NF-kappaB Non-Canonical Pathway Antibody Sampler Kit, 1 Kit
SDP
Catalog No. 502051208
Encompass
The NF-kappaB Non-Canonical Pathway Antibody Sampler Kit provides a comprehensive selection of antibodies for evaluating the activation state and total protein levels of key components involved in the noncanonical NF-κB signaling pathway. This kit is designed for sensitive detection of pathway regulators and signaling intermediates, including NIK, IKKα, TRAF2, TRAF3, and NF-κB2 p100/p52, using a variety of immunoassays.
  • Includes antibodies for monitoring pathway activation and protein expression, such as phospho-IKKα/β (Ser176/180) and phospho-NF-κB2 p100 (Ser866/870).
  • Contains secondary detection reagents for HRP-linked immunoblotting.
  • Validated for multiple applications, including Western blotting, immunoprecipitation, and immunohistochemistry.
  • Supplied in a stabilized glycerol buffer for storage at -20°C.
Catalog No. 50-205-1208 Supplier Cell Signaling Technology Supplier No. 4888T
The NF-kappaB Non-Canonical Pathway Antibody Sampler Kit provides a comprehensive selection of antibodies for evaluating the activation state and total protein levels of key components involved in the noncanonical NF-κB signaling pathway. This kit is designed for sensitive detection of pathway regulators and signaling intermediates, including NIK, IKKα, TRAF2, TRAF3, and NF-κB2 p100/p52, using a variety of immunoassays.
  • Includes antibodies for monitoring pathway activation and protein expression, such as phospho-IKKα/β (Ser176/180) and phospho-NF-κB2 p100 (Ser866/870).
  • Contains secondary detection reagents for HRP-linked immunoblotting.
  • Validated for multiple applications, including Western blotting, immunoprecipitation, and immunohistochemistry.
  • Supplied in a stabilized glycerol buffer for storage at -20°C.