Learn More
Cell Signaling Technology NF-kappaB Pathway Antibody Sampler Kit II, 1 Kit
SDP
Catalog No. 502732338
Encompass
The NF-κB Pathway Antibody Sampler Kit II provides a comprehensive set of reagents for investigating the activation state and total protein levels of key signaling molecules within the NF-κB pathway. The kit features monoclonal antibodies designed to detect IKKα, IKKβ, NF-κB p65/RelA, and IκBα to facilitate detailed analysis of pathway regulation and protein phosphorylation.
  • Includes reagents for measuring total and phosphorylated protein levels
  • Sufficient volume to perform two western blot experiments with each primary antibody
  • Validated for diverse applications including western blotting and immunoprecipitation
  • Supplied with HRP-linked secondary antibody for convenient detection
  • For research use only
Catalog No. 50-273-2338 Supplier Cell Signaling Technology Supplier No. 64662T
The NF-κB Pathway Antibody Sampler Kit II provides a comprehensive set of reagents for investigating the activation state and total protein levels of key signaling molecules within the NF-κB pathway. The kit features monoclonal antibodies designed to detect IKKα, IKKβ, NF-κB p65/RelA, and IκBα to facilitate detailed analysis of pathway regulation and protein phosphorylation.
  • Includes reagents for measuring total and phosphorylated protein levels
  • Sufficient volume to perform two western blot experiments with each primary antibody
  • Validated for diverse applications including western blotting and immunoprecipitation
  • Supplied with HRP-linked secondary antibody for convenient detection
  • For research use only