Learn More
Enzo Life Sciences NF-kappaB (p65) (CT) polyclonal antibody (100 µl)
SDP
Supplier:  Enzo Life Sciences BMLSA1710100
Encompass_Preferred
Immunogen: Synthetic peptide corresponding to a portion of C-terminal human NF-κB p65. Species reactivity: Human, Mouse. Specificity: Recognizes the p65 subunit of NFκB. Applications: ELISA, IP, WB EMSA. Formulation: Liquid. Neat serum.
Catalog No. 50-201-1763