Learn More
Abcam NF kappaB p65 ELISA Kit, 1 x 96 Tests
Catalog No. 502582294
Encompass_Preferred
NF kappaB p65 ELISA Kit is a single-wash 90-min Simplestep used to quantify NF kappaB p65 with a sensitivity of 5 µg/mL. The assay uses a simple mix-wash-read protocol with just one incubation and one wash step. - Colorimetric Sandwich ELISA - 450 nm readout : works on any standard plate reader - Cited in over 30 citations
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-258-2294 Supplier Abcam Supplier No. AB1766481X96TESTS
NF kappaB p65 ELISA Kit is a single-wash 90-min Simplestep used to quantify NF kappaB p65 with a sensitivity of 5 µg/mL. The assay uses a simple mix-wash-read protocol with just one incubation and one wash step. - Colorimetric Sandwich ELISA - 450 nm readout : works on any standard plate reader - Cited in over 30 citations
The product is subject to the following: Abcam Restricted Use Statement