Learn More
Enzo Life Sciences NF-kappaB p65 polyclonal antibody (200µg)
SDP
Supplier:  Enzo Life Sciences ADIKASTF110F
Encompass_Preferred
Host: Rabbit. Immunogen: Synthetic peptide corresponding to a portion of human p65 NF-κB. The sequence is completely conserved in mouse NF-κB p65. UniProt ID: Q04206. Source: Purified from rabbit serum. Species reactivity: Human, Mouse, Rat, Rabbit, Porcine, Monkey, Guinea pig, Dog, Bovine. Applications: WB. Recommended Dilutions: Western Blot (1:1,000, colorimetric) Suggested dilutions/conditions may not be available for all applications. Optimal conditions must be determined individually for each application. Purity: Protein A affinity purified. Formulation: Liquid. In PBS, pH 7.2, containing 0.09% sodium azide. Handling: Avoid freeze/thaw cycles.
Catalog No. 50-200-7323