Learn More
TARGETMOL CHEMICALS INC NF-KAPPABETA ACTIVATOR 1 5MG
Supplier: TARGETMOL CHEMICALS INC T399655MG
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. NF-(kappa)(beta) activator 1 can activate the nf-kappa (beta) gene activator EC50 of 0.9 microns. NF-(kappa)(beta) is widely used in eukaryotic cells as a gene regulating cell proliferation and cell survival. NF-(kappa)(beta) activator 1 induces superoxide dismutase (SOD) 2 1 mRNA expression. purity: 98%