Learn More
Cayman Chemical NF-KB InhIbItrtrIfluoroace 5mg
SDP
Catalog No. 502966140
Encompass_Preferred
A synthetic peptide corresponding to the nuclear localization sequence of NF-κB p105 subunit (also known as p50) appended to a hydrophobic sequence to facilitate import into living cells; blocks the nuclear import of p105 in cells treated with activators of NF-κB signaling, including LPS and TNF-α
Catalog No. 50-296-6140 Supplier Cayman Chemical Supplier No. 174935 MG
A synthetic peptide corresponding to the nuclear localization sequence of NF-κB p105 subunit (also known as p50) appended to a hydrophobic sequence to facilitate import into living cells; blocks the nuclear import of p105 in cells treated with activators of NF-κB signaling, including LPS and TNF-α