Learn More
BPS Bioscience Inc NF-κB Luciferase Reporter Lentivirus, Firefly Luciferase, Lentivirus, VSV-G pseudotyped, Replication incompetent, Shelf Life 12 months, BSL-2, Titer ≥10^7 TU/ml, Volume 500 μl x 2
SDP
Catalog No. NC1992967
Encompass
NF-κB Luciferase Reporter Lentivirus, Firefly Luciferase, Lentivirus, VSV-G pseudotyped, Replication incompetent, Shelf Life 12 months, BSL-2, Titer ≥10^7 TU/ml, Volume 500 μl x 2
Catalog No. NC1992967 Supplier BPS Bioscience Inc Supplier No. 79564
NF-κB Luciferase Reporter Lentivirus, Firefly Luciferase, Lentivirus, VSV-G pseudotyped, Replication incompetent, Shelf Life 12 months, BSL-2, Titer ≥10^7 TU/ml, Volume 500 μl x 2