Learn More
AdipoGen NF-kB p65 pAb
SDP
Supplier:  AdipoGen AG25T0004R100
Encompass_Preferred
Polyclonal Antibody. Recognizes human and mouse NF-kappaB p65 subunit. Source Rabbit. Applications ICC, IP, WB. Liquid. Contains 0.1% sodium azide. NF-kappaB is a pleiotropic transcription factor present in almost all cell types and is the endpoint of a series of signal transduction events that are initiated by a vast array of stimuli related to many biological processes such as inflammation, immunity, differentiation, cell growth, tumorigenesis and apoptosis. NF-kappaB is a homo- or heterodimeric complex formed by the Rel-like domain-containing proteins RELA/p65, RELB, NFKB1/p105, NFKB1/p50, REL and NFKB2/p52. The heterodimeric p65-p50 complex is the most abundant complex. The dimers bind at kappa-B sites in the DNA of their target genes and the individual dimers have distinct preferences for different kappaB sites that they can bind with distinguishable affinity and specificity. Different dimer combinations act as transcriptional activators or repressors, respectively.
Catalog No. 50-256-3174