Learn More
Apexbio Technology LLC DiscoveryProbe™ NF-B Signaling Compound Library Plus, 100uL/well(10 mM solution), 96 Well Racks With Screw Cap
SDP
Supplier:  Apexbio Technology LLC L1044P.1
Encompass_Preferred
DiscoveryProbe™ NF-B Signaling Compound Library Plus is a unique collection of 178 NF-B inhibitors for NF-B signaling pathway research. Available in stock with overnight delivery and free shipping over $500 Cost-effective and competitive price to save your findings Potent, selective and cell-permeable in inhibiting or activating target molecules Diverse in chemical structure and route of administration (oral/i.m/i.v injection etc.) Detailed files describing potency, selectivity and applications etc. Supported by published data from top peer-reviewed journals Guaranteed high quality with NMR and HPLC validation. Other sizes are also available. Please inqury us for quote.
Catalog No. 50-199-8485