Learn More
Enzo Life Sciences Neurofilament triplet proteins monoclonal antibody (13AA) (100µl)
SDP
Supplier:  Enzo Life Sciences BMLNA12230100
Encompass_Preferred
Clone: 13AA. Isotype: IgG. Immunogen: Purified human spinal cord neurofilament triplet proteins. Species reactivity: Human, Mouse, Rat Mammalian. Applications: IHC, WB. Purity: Chromatographically purified. Formulation: Liquid. In buffer.
Catalog No. 50-201-1178