Learn More
Medchemexpress LLC NF023 hexasodium | 104869-31-0 | MFCD01074985 | 100.0% | 1162.88 g·mol⁻¹ | C35H20N4Na6O21S6 | 5 MG
SDP
Catalog No. 5000263474
Encompass_Preferred
NF023 hexasodium is the hexasodium salt of NF023, a selective and competitive antagonist of the P2X1 receptor used in research applications. It exhibits an IC50 of 0.21 μM for human P2X1 and markedly reduced activity at other P2X subtypes. Supplied as a high-purity dry solid in milligram quantities for laboratory assays and screening; for research use only.
  • Selective P2X1 receptor antagonist with reported IC50 of 0.21 μM.
  • High purity to support experimental reproducibility.
  • Available in small milligram sizes suitable for in vitro studies.
  • Provided as the hexasodium salt with defined molecular weight and formula.
  • For research use only; not for human or veterinary use.
Catalog No. 50-002-63474 Supplier Medchemexpress LLC Supplier No. HY1086765MG
NF023 hexasodium is the hexasodium salt of NF023, a selective and competitive antagonist of the P2X1 receptor used in research applications. It exhibits an IC50 of 0.21 μM for human P2X1 and markedly reduced activity at other P2X subtypes. Supplied as a high-purity dry solid in milligram quantities for laboratory assays and screening; for research use only.
  • Selective P2X1 receptor antagonist with reported IC50 of 0.21 μM.
  • High purity to support experimental reproducibility.
  • Available in small milligram sizes suitable for in vitro studies.
  • Provided as the hexasodium salt with defined molecular weight and formula.
  • For research use only; not for human or veterinary use.