Learn More
Cayman Chemical NF449 (sodium salt), Purity: ≥95%, CAS: 627034-85-9, 50 mg
SDP
Catalog No. 502952975
Encompass_Preferred
An analog of suramin that selectively inhibits P2X1 receptors (pIC50 = 6.3); selectively antagonizes the Gsα-subunit G protein, which suppresses the association rate of GTPγS binding to Gsα-s, inhibits the stimulation of adenylyl cyclase activity, and blocks G protein coupling to certain GPCRs
Catalog No. 50-295-2975 Supplier Cayman Chemical Supplier No. 1332450 MG
An analog of suramin that selectively inhibits P2X1 receptors (pIC50 = 6.3); selectively antagonizes the Gsα-subunit G protein, which suppresses the association rate of GTPγS binding to Gsα-s, inhibits the stimulation of adenylyl cyclase activity, and blocks G protein coupling to certain GPCRs