Learn More
Sino Biological NFASC / Neurofascin Antibody, Rabbit PAb, Antigen Affinity Purified 100uL
Supplier:  Sino Biological 15694T16100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Human NFASC / Neurofascin (rh NFASC / Neurofascin; Catalog#15694-H08H; O94856-12; Met1-Gln939). NFASC / Neurofascin specific IgG was purified by Human NFASC / Neurofascin affinity chromatography.
Catalog No. 50-224-8771