Learn More
Cayman Chemical NFAT Activation Inhibitor III, Purity: ≥98%, CAS: 3519-82-2, 25 mg
SDP
Catalog No. 502986654
Encompass_Preferred
A cell-permeant inhibitor of NFAT signaling; binds calcineurin (Kd = 0.8 µM), preventing it from dephosphorylating NFAT
Catalog No. 50-298-6654 Supplier Cayman Chemical Supplier No. 2181225 MG
A cell-permeant inhibitor of NFAT signaling; binds calcineurin (Kd = 0.8 µM), preventing it from dephosphorylating NFAT