Learn More
Cayman Chemical NFAT AcTVatIon InHBTr III 50mg
SDP
Catalog No. 502986099
Encompass_Preferred
A cell-permeant inhibitor of NFAT signaling; binds calcineurin (Kd = 0.8 µM), preventing it from dephosphorylating NFAT
Catalog No. 50-298-6099 Supplier Cayman Chemical Supplier No. 2181250 MG
Add to Cart
Add to Cart
A cell-permeant inhibitor of NFAT signaling; binds calcineurin (Kd = 0.8 µM), preventing it from dephosphorylating NFAT