Learn More
BPS Bioscience Inc NFAT EGFP REPORTER LENTIVIRUS
SDP
Catalog No. NC1920699
Encompass
The NFAT eGFP Reporter Lentivirus are replication incompetent, HIV-based, VSV-G pseudotyped lentiviral particles that are ready to be transduced into almost all types of mammalian cells, including primary and non-dividing cells. BPS BIOSCIENCE INC mfg# 79922
Catalog No. NC1920699 Supplier BPS Bioscience Inc Supplier No. 79922
The NFAT eGFP Reporter Lentivirus are replication incompetent, HIV-based, VSV-G pseudotyped lentiviral particles that are ready to be transduced into almost all types of mammalian cells, including primary and non-dividing cells. BPS BIOSCIENCE INC mfg# 79922