Learn More
Medchemexpress LLC NFAT Inhibitor-1 (VIVIT peptide) | 249537-73-3 | 99.9% | 1683.93 | 25 MG
SDP
Catalog No. 5000425452
Encompass_Preferred
NFAT Inhibitor-1 (VIVIT peptide) is a potent inhibitor of nuclear factor of activated T-cells (NFAT). It selectively inhibits calcineurin-mediated dephosphorylation of NFAT, making it suitable for research applications.
  • Selectively inhibits calcineurin-mediated dephosphorylation of NFAT
  • Significantly inhibits nuclear translocation of NFATc1
  • Long-term treatment inhibits cytoplasmic levels of cathepsin K, TRAP, and MMP-9
  • Inhibits protein expression of NFATC2 in hLESCs
  • Inhibits the effect of WNT16b on cell proliferation in hLESCs
  • Significantly inhibits expression of WNT16b-promoting cell proliferation marker Ki67 and the putative hLESC marker ΔNp63α in hLESCs
Catalog No. 50-004-25452 Supplier Medchemexpress LLC Supplier No. 25MGHYP1026
NFAT Inhibitor-1 (VIVIT peptide) is a potent inhibitor of nuclear factor of activated T-cells (NFAT). It selectively inhibits calcineurin-mediated dephosphorylation of NFAT, making it suitable for research applications.
  • Selectively inhibits calcineurin-mediated dephosphorylation of NFAT
  • Significantly inhibits nuclear translocation of NFATc1
  • Long-term treatment inhibits cytoplasmic levels of cathepsin K, TRAP, and MMP-9
  • Inhibits protein expression of NFATC2 in hLESCs
  • Inhibits the effect of WNT16b on cell proliferation in hLESCs
  • Significantly inhibits expression of WNT16b-promoting cell proliferation marker Ki67 and the putative hLESC marker ΔNp63α in hLESCs