Learn More
Medchemexpress LLC NFAT Inhibitor-1 (VIVIT peptide) | 249537-73-3 | 99.9% | 1683.93 | 5 MG
SDP
Catalog No. 5000424906
Encompass_Preferred
NFAT Inhibitor-1 (VIVIT peptide) is an inhibitor of nuclear factor of activated Tcells (NFAT) that selectively inhibits calcineurin-mediated dephosphorylation of NFAT. It is for research use only and not sold to patients.
  • Selectively inhibits calcineurin-mediated dephosphorylation of NFAT
  • Significantly inhibits nuclear translocation of NFATc1 for 24 hours
  • Long-term treatment significantly inhibits the cytoplasmic levels of cathepsin K, TRAP, and MMP-9
Catalog No. 50-004-24906 Supplier Medchemexpress LLC Supplier No. HYP10265MG
Add to Cart
Add to Cart
NFAT Inhibitor-1 (VIVIT peptide) is an inhibitor of nuclear factor of activated Tcells (NFAT) that selectively inhibits calcineurin-mediated dephosphorylation of NFAT. It is for research use only and not sold to patients.
  • Selectively inhibits calcineurin-mediated dephosphorylation of NFAT
  • Significantly inhibits nuclear translocation of NFATc1 for 24 hours
  • Long-term treatment significantly inhibits the cytoplasmic levels of cathepsin K, TRAP, and MMP-9